2,4-Diaminopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: Pyrimidine-2,4-diamine, Pyrimidine-2,4-diyltriamine

A508788-100G A508788-1G A508788-5G
77292-170EA 1024.85 USD
77292-170 77292-172 77292-174
2,4-Diaminopyrimidine 98%
2,4-Diaminopyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD00038023
CAS Number: 156-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR