2-(4-Chlorophenyl)ethyl chloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-(2-chloroethyl)benzene

A508768-1G A508768-5G
77241-580EA 56.65 USD
77241-580 77241-582
2-(4-Chlorophenyl)ethyl chloride 97%
2-(4-Chlorophenyl)ethyl chloride

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.06 g/mol
MDL Number: MFCD00045294
CAS Number: 32327-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR