Flupirtine maleate salt 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl (2-amino-6-((4-fluorobenzyl)amino)pyridin-3-yl)carbamate maleate, 2-Amino-3-(ethoxycarbonylamino)-6-(4-fluorobenzylamino)pyridine Maleate Salt, Flupirtine Maleate

A508491-100MG A508491-10MG A508491-1MG A508491-250MG A508491-50MG
77253-378EA 19 USD
77253-378 77253-380 77253-382 77253-384 77253-386
Flupirtine maleate salt 98%
Flupirtine maleate salt

Precaución: for Research Use Only

Formula: C₁₅H₁₇FN₄O₂
MW: 420.40 g/mol
MDL Number: MFCD00941415
CAS Number: 75507-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR