(±)-1-Octyn-3-ol 98%

Supplier: Ambeed
Danger

Synonyms: Oct-1-yn-3-ol

A507946-100G A507946-25G A507946-5G
77241-560EA 200.85 USD
77241-560 77241-562 77241-564
(±)-1-Octyn-3-ol 98%
(±)-1-Octyn-3-ol

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00004588
CAS Number: 818-72-4
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR