Cis-methyl 2-hydroxycyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A507907-100MG A507907-1G A507907-250MG
77265-754EA 63.86 USD
77265-754 77265-756 77265-758
Cis-methyl 2-hydroxycyclohexanecarboxylate 97%
Cis-methyl 2-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.19 g/mol
MDL Number: MFCD00064948
CAS Number: 936-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR