4-Phenylthiophene-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A507290-1G A507290-250MG A507290-5G
77292-150EA 32 USD
77292-150 77292-152 77292-154
4-Phenylthiophene-2-carbaldehyde 97%
4-Phenylthiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈OS
MDL Number: MFCD03701610
CAS Number: 26170-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR