4,4'-(Anthracene-9,10-diyl)dibenzaldehyde 98%

Supplier: Ambeed
Warning

A507266-100MG A507266-250MG
77253-366EA 42.23 USD
77253-366 77253-368
4,4'-(Anthracene-9,10-diyl)dibenzaldehyde 98%
4,4'-(Anthracene-9,10-diyl)dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₈H₁₈O₂
MW: 386.44 g/mol
MDL Number: MFCD18968034
CAS Number: 324750-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR