1-(2-Hydroxyethyl)-2,3,3-trimethyl-3H-indol-1-ium bromide 97%

Supplier: Ambeed
Warning

A507051-1G A507051-250MG A507051-5G
77265-730EA 64 USD
77265-730 77265-732 77265-734
1-(2-Hydroxyethyl)-2,3,3-trimethyl-3H-indol-1-ium bromide 97%
1-(2-Hydroxyethyl)-2,3,3-trimethyl-3H-indol-1-ium bromide

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO
MW: 284.19 g/mol
MDL Number: MFCD00980427
CAS Number: 29636-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR