4-Bromo-2-methylquinoline 97%

Supplier: Ambeed
Warning

A507021-1G A507021-250MG A507021-5G
77292-142EA 21.63 USD
77292-142 77292-144 77292-146 77292-148
4-Bromo-2-methylquinoline 97%
4-Bromo-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD02278398
CAS Number: 50488-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR