7-(Difluoromethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A507002-100MG
A507002-1G
A507002-250MG
77241-554EA
42.23
USD
77241-554
77241-556
77241-558
7-(Difluoromethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine 97%
7-(Difluoromethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine
Precaución: for Research Use Only
Formula:
C₉H₉F₂N₃ MW: 197.18 g/mol |
MDL Number:
MFCD28969780 CAS Number: 1795186-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...