(2R,4aR,6S,7R,8S,8aR)-6-(Ethylthio)-8-(naphthalen-2-ylmethoxy)-2-phenylhexahydropyrano[3,2-d][1,3]dioxin-7-yl benzoate 98%

Supplier: Ambeed
Warning

A506940-100MG A506940-1G A506940-250MG
77265-724EA 28 USD
77265-724 77265-726 77265-728
(2R,4aR,6S,7R,8S,8aR)-6-(Ethylthio)-8-(naphthalen-2-ylmethoxy)-2-phenylhexahydropyrano[3,2-d][1,3]dioxin-7-yl benzoate 98%
(2R,4aR,6S,7R,8S,8aR)-6-(Ethylthio)-8-(naphthalen-2-ylmethoxy)-2-phenylhexahydropyrano[3,2-d][1,3]dioxin-7-yl benzoate

Precaución: for Research Use Only

Formula: C₃₃H₃₂O₆S
MDL Number: MFCD13182917
CAS Number: 352008-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR