4-(Difluoromethoxy)benzylamine 95%

Supplier: Ambeed
Danger

Synonyms: (4-(Difluoromethoxy)phenyl)methanamine

A506631-250MG A506631-5G A506631-1G
77292-134EA 12 USD
77292-134 77292-136 77749-032
4-(Difluoromethoxy)benzylamine 95%
4-(Difluoromethoxy)benzylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂NO
MDL Number: MFCD00236231
CAS Number: 177842-14-7
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR