4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(4,4'-Biphenyldiyl)bis(1H-imidazole)

A506472-250MG A506472-5G
77241-550EA 36.05 USD
77241-550 77241-552
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl 98%
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₄N₄
MW: 286.34 g/mol
MDL Number: MFCD23103195
CAS Number: 855766-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR