2,3,5-Trichloro-pyridin-4-ol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3,5-Trichloropyridin-4-ol
A506320-1G
A506320-250MG
A506320-100MG
77253-358EA
363.59
USD
77253-358
77253-360
77747-040
2,3,5-Trichloro-pyridin-4-ol 95%
2,3,5-Trichloro-pyridin-4-ol
Precaución: for Research Use Only
Formula:
C₅H₂Cl₃NO MW: 198.434 g/mol |
MDL Number:
MFCD01690819 CAS Number: 1970-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...