5-Bromo-7-methylbenzo[b]thiophene 97%

Supplier: Ambeed
Warning

A506207-250MG A506207-5G
77241-546EA 212.18 USD
77241-546 77241-548
5-Bromo-7-methylbenzo[b]thiophene 97%
5-Bromo-7-methylbenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₉H₇BrS
MW: 227.12 g/mol
MDL Number: MFCD22560426
CAS Number: 19076-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR