4-(Ethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(N-Ethylaminocarbonyl)phenylboronic acid

A505532-1G A505532-250MG
77292-130EA 106.09 USD
77292-130 77292-132
4-(Ethylcarbamoyl)phenylboronic acid 98%
4-(Ethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₃
MW: 193.0100 g/mol
MDL Number: MFCD03411954
CAS Number: 850568-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR