3,5-Lutidine-N-oxide 98%

Supplier: Ambeed
Warning

A505470-1G A505470-25G A505470-5G A505470-100G
77279-710EA 22 USD
77279-710 77279-712 77279-714 77749-030
3,5-Lutidine-N-oxide 98%
3,5-Lutidine N-oxide

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD00126839
CAS Number: 3718-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR