5-Hydroxy-2-methylindole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1H-indol-5-ol

A505321-1G A505321-250MG A505321-5G
77279-704EA 15 USD
77279-704 77279-706 77279-708
5-Hydroxy-2-methylindole 97%
5-Hydroxy-2-methylindole

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00022714
CAS Number: 13314-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR