4-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3-(trifluoromethyl)phenyl)methanol

A504843-1G A504843-25G A504843-5G
77241-540EA 27 USD
77241-540 77241-542 77241-544
4-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%
4-Fluoro-3-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD00061171
CAS Number: 67515-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR