2,4,5-Tribromo-1-methylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,5-Tribromo-1-methyl-1H-imidazole

A504211-100G A504211-1G A504211-25G A504211-5G A504211-10G
77253-346EA 449.08 USD
77253-346 77253-348 77253-350 77253-352 77747-035
2,4,5-Tribromo-1-methylimidazole 98%
2,4,5-Tribromo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₃Br₃N₂
MDL Number: MFCD00955564
CAS Number: 1003-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR