1,1'-((((5-Chloropyrimidine-2,4-diyl)bis(azanediyl))bis(3-methoxy-4,1-phenylene))bis(piperazine-4,1-diyl))bis(ethan-1-one) 98+%

Supplier: Ambeed
Warning

A503888-10MG A503888-1MG A503888-25MG A503888-50MG A503888-5MG
77241-528EA 49 USD
77241-528 77241-530 77241-532 77241-534 77241-536
1,1'-((((5-Chloropyrimidine-2,4-diyl)bis(azanediyl))bis(3-methoxy-4,1-phenylene))bis(piperazine-4,1-diyl))bis(ethan-1-one) 98+%
1,1'-((((5-Chloropyrimidine-2,4-diyl)bis(azanediyl))bis(3-methoxy-4,1-phenylene))bis(piperazine-4,1-diyl))bis(ethan-1-one)

Precaución: for Research Use Only

Formula: C₃₀H₃₇ClN₈O₄
MDL Number: MFCD28133395
CAS Number: 1472795-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR