(2-Chloro-4-methoxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-4-methoxyphenyl)acetic acid

A503779-1G A503779-250MG
77265-700EA 113.3 USD
77265-700 77265-702
(2-Chloro-4-methoxyphenyl)acetic acid 95%
(2-Chloro-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
MDL Number: MFCD02664702
CAS Number: 91367-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR