4-Bromo-3,5-dihydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A503597-1G A503597-25G A503597-5G
77279-698EA 5.15 USD
77279-698 77279-700 77279-702
4-Bromo-3,5-dihydroxybenzoic acid 95%
4-Bromo-3,5-dihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₄
MW: 233.02 g/mol
MDL Number: MFCD00002513
CAS Number: 16534-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR