rel-2,7-Bis(((3R,5S)-3,5-dimethylpiperidin-1-yl)sulfonyl)anthracene-9,10-dione dioxime 98%

Supplier: Ambeed
Danger

Synonyms: Tegatrabetan

A503122-10MG A503122-1MG A503122-5MG A503122-100MG A503122-50MG
77265-694EA 98.88 USD
77265-694 77265-696 77265-698 77617-150 77617-152
rel-2,7-Bis(((3R,5S)-3,5-dimethylpiperidin-1-yl)sulfonyl)anthracene-9,10-dione dioxime 98%
rel-2,7-Bis(((3R,5S)-3,5-dimethylpiperidin-1-yl)sulfonyl)anthracene-9,10-dione dioxime

Precaución: for Research Use Only

Formula: C₂₈H₃₆N₄O₆S₂
MW: 588.74 g/mol
MDL Number: MFCD32633578
CAS Number: 1227637-23-1
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR