(±)-4-Hydroxy-3-methoxymandelic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-methoxy-DL-mandelic acid, VMA, Vanillyl mandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, (±)-4-Hydroxy-3-methoxyphenylglycolic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, Vanillylmandelic acid

A503085-5G A503085-250MG A503085-1G
77253-328EA 151 USD
77253-328 77572-604 77747-032
(±)-4-Hydroxy-3-methoxymandelic acid 97%
(±)-4-Hydroxy-3-methoxymandelic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 421
MDL Number: MFCD00004235
CAS Number: 55-10-7
Merck Index: 12,10070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR