3-(5-(4-(1H-Imidazol-1-yl)phenyl)-1-(4-carbamoyl-2-methylphenyl)-1H-pyrrol-2-yl)propanoic acid 98+%

Supplier: Ambeed
Warning

A502981-10MG A502981-1MG A502981-50MG A502981-5MG
77241-522EA 50 USD
77241-522 77241-524 77241-526 77617-148
3-(5-(4-(1H-Imidazol-1-yl)phenyl)-1-(4-carbamoyl-2-methylphenyl)-1H-pyrrol-2-yl)propanoic acid 98+%
3-(5-(4-(1H-Imidazol-1-yl)phenyl)-1-(4-carbamoyl-2-methylphenyl)-1H-pyrrol-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₄O₃
MDL Number: MFCD22123247
CAS Number: 1208315-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR