2-Amino-5-bromo-4-fluorobenzamide 97%

Supplier: Ambeed
Warning

A502628-100MG A502628-1G A502628-250MG
77241-516EA 8 USD
77241-516 77241-518 77241-520
2-Amino-5-bromo-4-fluorobenzamide 97%
2-Amino-5-bromo-4-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₆BrFN₂O
MDL Number: MFCD14585387
CAS Number: 1206969-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR