ARS-853 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-(4-((4-Chloro-2-hydroxy-5-(1-methylcyclopropyl)phenyl)glycyl)piperazin-1-yl)azetidin-1-yl)prop-2-en-1-one

A502082-1MG A502082-25MG A502082-5MG A502082-50MG A502082-10MG
77265-682EA 53.56 USD
77265-682 77265-684 77265-686 77749-025 77749-024
ARS-853 98%
ARS-853

Precaución: for Research Use Only

Formula: C₂₂H₂₉ClN₄O₃
MW: 432.94 g/mol
MDL Number: MFCD30532658
CAS Number: 1629268-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR