Ethyl 3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-β-D-thioglucopyranoside 98%

Supplier: Ambeed
Warning

A502030-1G A502030-250MG A502030-5G
77253-316EA 32.96 USD
77253-316 77253-318 77253-320
Ethyl 3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-β-D-thioglucopyranoside 98%
Ethyl 3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-β-D-thioglucopyranoside

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₉S
MW: 479.50 g/mol
MDL Number: MFCD15144989
CAS Number: 99409-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR