(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethyldi(3,5-xylyl)phosphine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A501718-1G
A501718-100MG
77253-308EA
403.76
USD
77253-308
77747-028
(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethyldi(3,5-xylyl)phosphine ≥98%, ee 99%
(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethyldi(3,5-xylyl)phosphine
Precaución: for Research Use Only
Formula:
C₄₆H₅₂FeO₂P₂ MW: 754.70 g/mol |
MDL Number:
MFCD08561165 CAS Number: 849924-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...