3-Acetylbenzoic acid 98%

Supplier: Ambeed
Warning

A501579-1G A501579-5G A501579-25G
77265-678EA 12 USD
77265-678 77265-680 77572-590
3-Acetylbenzoic acid 98%
3-Acetylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00045847
CAS Number: 586-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR