4-Bromo-N,N-diethylbenzamide 95%

Supplier: Ambeed
Warning

A500655-1G A500655-25G A500655-5G
77292-112EA 11 USD
77292-112 77292-114 77292-116
4-Bromo-N,N-diethylbenzamide 95%
4-Bromo-N,N-diethylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
MW: 256.1390 g/mol
MDL Number: MFCD00463697
CAS Number: 5892-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR