(2-Aminophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A500358-1G A500358-250MG A500358-25G A500358-5G
77253-270EA 83 USD
77253-270 77253-272 77253-274 77253-276
(2-Aminophenyl)acetic acid 95%
(2-Aminophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD03701521
CAS Number: 3342-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR