3-Bromo-2-chloro-4-(difluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A500322-1G A500322-250MG
77265-666EA 260 USD
77265-666 77265-668
3-Bromo-2-chloro-4-(difluoromethoxy)pyridine 95%
3-Bromo-2-chloro-4-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClF₂NO
MDL Number: MFCD28733844
CAS Number: 1805137-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR