3-Chloro-4-hydroxyphenylacetic acid 95%

Supplier: Ambeed
Warning

A500222-25G A500222-5G
77241-506EA 71.07 USD
77241-506 77241-508
3-Chloro-4-hydroxyphenylacetic acid 95%
3-Chloro-4-hydroxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00004349
CAS Number: 33697-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR