2-Trimethylsilylthiophene 98+%

Supplier: Ambeed
Danger

Synonyms: 2-Thienyltrimethylsilane, Trimethyl(thiophen-2-yl)silane

A499679-25G A499679-5G
77279-680EA 94.76 USD
77279-680 77279-682
2-Trimethylsilylthiophene 98+%
2-Trimethylsilylthiophene

Precaución: for Research Use Only

Formula: C₇H₁₂SSi
MW: 156.32 g/mol
MDL Number: MFCD00005415
CAS Number: 18245-28-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR