2-(Bromomethyl)-5-chlorobenzonitrile 95%

Supplier: Ambeed
Danger

A499626-1G A499626-250MG A499626-5G
77241-500EA 83 USD
77241-500 77241-502 77241-504
2-(Bromomethyl)-5-chlorobenzonitrile 95%
2-(Bromomethyl)-5-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrClN
MDL Number: MFCD09909826
CAS Number: 50712-69-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR