1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%

Supplier: Ambeed
Warning

A499566-100MG A499566-1G A499566-250MG
77253-262EA 145 USD
77253-262 77253-264 77253-266
1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I)

Precaución: for Research Use Only

Formula: C₃₀H₃₄FeP₂Rh
MDL Number: MFCD17018836
CAS Number: 854275-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR