2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-6-sulphonyl chloride

A499491-1G A499491-250MG A499491-5G
77241-494EA 13 USD
77241-494 77241-496 77241-498
2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride 98%
2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₇ClO₄S
MDL Number: MFCD02677681
CAS Number: 63758-12-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR