3-Methoxy-4-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxy-4-methylbenzeneboronic acid

A499234-1G A499234-5G
77279-678EA 31.93 USD
77279-678 77749-023
3-Methoxy-4-methylphenylboronic acid 98%
3-Methoxy-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
MDL Number: MFCD08274479
CAS Number: 917757-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR