Ax20017 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclopropanecarboxamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide

A499106-10MG A499106-50MG A499106-5MG A499106-1MG
77265-652EA 99.91 USD
77265-652 77265-654 77265-656 77749-022
Ax20017 98%
Ax20017

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂S
MW: 264.34 g/mol
MDL Number: MFCD00785286
CAS Number: 329221-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR