2-(Methylthio)benzaldehyde 98%

Supplier: Ambeed
Warning

A498934-1G A498934-250MG A498934-5G
77253-256EA 28 USD
77253-256 77253-258 77253-260
2-(Methylthio)benzaldehyde 98%
2-(Methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈OS
MDL Number: MFCD00196822
CAS Number: 7022-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR