Chlorogold,[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane 98%

Supplier: Ambeed
Warning

A498530-100MG A498530-1G A498530-250MG
77253-250EA 60 USD
77253-250 77253-252 77253-254
Chlorogold,[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane 98%
Chlorogold,[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane

Precaución: for Research Use Only

Formula: C₄₄H₃₂AuClP₂
MDL Number: MFCD15144870
CAS Number: 685138-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR