Methyl 5-fluoro-2-methylnicotinate 97%

Supplier: Ambeed
Warning

A498507-100MG A498507-250MG A498507-5G
77253-244EA 94.76 USD
77253-244 77253-246 77253-248
Methyl 5-fluoro-2-methylnicotinate 97%
Methyl 5-fluoro-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD18254252
CAS Number: 868636-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR