Methyl 4-bromo-2-(bromomethyl)-6-fluorobenzoate 97%

Supplier: Ambeed
Warning

A498492-100MG A498492-1G A498492-250MG
77265-640EA 45 USD
77265-640 77265-642 77265-644
Methyl 4-bromo-2-(bromomethyl)-6-fluorobenzoate 97%

Precaución: for Research Use Only

Formula: C₉H₇Br₂FO₂
MDL Number: MFCD29760551
CAS Number: 1935921-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR