(S)-5-((3-Phenoxybenzyl)(1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)benzene-1,2,4-tricarboxylic acid 98%

Supplier: Ambeed
Warning

A497149-10MG A497149-1MG A497149-50MG A497149-5MG
77241-474EA 39 USD
77241-474 77241-476 77241-478 77241-480
(S)-5-((3-Phenoxybenzyl)(1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)benzene-1,2,4-tricarboxylic acid 98%
(S)-5-((3-Phenoxybenzyl)(1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)benzene-1,2,4-tricarboxylic acid

Precaución: for Research Use Only

Formula: C₃₃H₂₇NO₈
MDL Number: MFCD28044314
CAS Number: 475205-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR