2,5-Bis(hexyl)-1,4-dibromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dibromo-2,5-dihexylbenzene

A497021-10G A497021-1G A497021-250MG A497021-5G
77253-234EA 262.99 USD
77253-234 77253-236 77253-238 77253-240
2,5-Bis(hexyl)-1,4-dibromobenzene 98%
2,5-Bis(hexyl)-1,4-dibromobenzene

Precaución: for Research Use Only

Formula: C₁₈H₂₈Br₂
MW: 404.24 g/mol
MDL Number: MFCD01318993
CAS Number: 117635-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR