2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture) 98% (mixture of cis and trans)

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-4-propyl-1,3-oxathiane

A496800-5G A496800-25G
77279-666EA 27.81 USD
77279-666 77749-016
2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture) 98% (mixture of cis and trans)
2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₈H₁₆OS
MW: 160.28 g/mol
MDL Number: MFCD00673203
CAS Number: 67715-80-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR