3-(4-Hydroxyphenyl)benzonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4'-Hydroxy-[1,1'-biphenyl]-3-carbonitrile, 4-(3-Cyanophenyl)phenol

A496784-100MG A496784-1G A496784-250MG A496784-5G
77292-092EA 29.87 USD
77292-092 77292-094 77292-096 77292-098
3-(4-Hydroxyphenyl)benzonitrile 98%
3-(4-Hydroxyphenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₉NO
MW: 195.2200 g/mol
MDL Number: MFCD05854255
CAS Number: 154848-44-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR