2-Amino-4-iodo-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A496755-100MG A496755-1G A496755-250MG
77265-624EA 65 USD
77265-624 77265-626 77265-628
2-Amino-4-iodo-5-(trifluoromethyl)pyridine 97%
2-Amino-4-iodo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃IN₂
MDL Number: MFCD16606194
CAS Number: 1227515-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR